Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
CONT 200ml PP TAPPO VITE ROSSO SING IR 1 * 150 pezzi
ANIC70.60.18ECSPI
:
Anicrin
CONT 200ml PP TAPPO VITE ROSSO SING IR 1 * 150 pezzi
ANIC70.60.18ECSPI
:
Anicrin
ANIC70.60.18ECSPI_2500050.pdf
CoC_ANIC70.60.18ECSPI_2100354001
CoC_ANIC70.60.18ECSPI_2200267
CoC_ANIC70.60.18ECSPI_2200803
CoC_ANIC70.60.18ECSPI_2200896
Frequently Bought Together
VWR®, Conductivity standards, Certified Reference Materials (CRMs) for primary calibration of conductivity meters
: VWR Chemicals
VWR®, Pipette sierologiche, Standard Line
: VWR Collection
Provette per centrifuga, Falcon®
: Corning
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Contenitori per campioni dritti con tappo a vite
: VWR Collection
VWR®, Piastra Petri, PS
: VWR Collection
VWR®, Provette e strip per PCR, 0,2 ml
: VWR Collection
pH electrodes, SenTix®
: WTW
VWR®, Provette per centrifuga, senza metalli, con tappo a vite
: VWR Collection
VWR®, Pipette Pasteur, Standard Line
: VWR Collection
VWR®, pipette di trasferimento, alte prestazioni
: VWR Collection
Provette per microcentrifuga Eppendorf® Protein LoBind®
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF