Orders LinkContactUsLinkComponent
Computers for Laboratory Equipment
Computers for Laboratory Equipment
Dell™ mouse, bundled with Dell™ keyboard, PT
Codice catalogo 906-0217
Nome del fornitore:   COLLECTION PREFIX HPLC
Computers for Laboratory Equipment
Codice catalogo 906-0217
Nome del fornitore:  COLLECTION PREFIX HPLC
Computers for Laboratory Equipment
Computers for Laboratory Equipment
Dell™ mouse, bundled with Dell™ keyboard, PT
Price
31.40 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
PC equipment optimised for use with laboratory devices and software. Designed to meet laboratory conditions, ensuring necessary performance. Includes a wide range of accessories such as monitors and keyboards.
  • Pre-installed with Windows LTSC 10 or 11 to reduce downtime from frequent updates and increase stability
  • Compact footprint with integrated PC-monitor solutions
  • Pre-installed language packs
  • Keyboards with all common European language layouts
  • Compatible with main CDS (Chromatography Data Systems)
Specifiche
  • Descrizione:
    Dell™ mouse, bundled with Dell™ keyboard, PT
  • Seq:
    28
Complementary Products
Informazioni sul prodotto
  • Codice catalogo
    Descrizione
    Disponibilità
    Price
  • 906-0219
    Dell™ mouse, bundled with Dell™ keyboard, DE
    Log in per disponibilità
    1 * 1 ST - SFr. 32.40
  • 906-0211
    Dell™ mouse, bundled with Dell™ keyboard, FR
    Log in per disponibilità
    1 * 1 ST - SFr. 32.40