Computers for Laboratory Equipment
Dell™ mouse, bundled with Dell™ keyboard, international
Codice catalogo 906-0224
Nome del fornitore: COLLECTION PREFIX HPLC
Computers for Laboratory Equipment
Codice catalogo 906-0224
Nome del fornitore: COLLECTION PREFIX HPLC
Product Details & Documents
PC equipment optimised for use with laboratory devices and software. Designed to meet laboratory conditions, ensuring necessary performance. Includes a wide range of accessories such as monitors and keyboards.
- Pre-installed with Windows LTSC 10 or 11 to reduce downtime from frequent updates and increase stability
- Compact footprint with integrated PC-monitor solutions
- Pre-installed language packs
- Keyboards with all common European language layouts
- Compatible with main CDS (Chromatography Data Systems)
Specifiche
- Descrizione:Dell™ mouse, bundled with Dell™ keyboard, international
- Seq:25