Computers for Laboratory Equipment
Dell™ mouse, bundled with Dell™ keyboard, DK
906-0215
PC equipment optimised for use with laboratory devices and software. Designed to meet laboratory conditions, ensuring necessary performance. Includes a wide range of accessories such as monitors and keyboards.
- Pre-installed with Windows LTSC 10 or 11 to reduce downtime from frequent updates and increase stability
- Compact footprint with integrated PC-monitor solutions
- Pre-installed language packs
- Keyboards with all common European language layouts
- Compatible with main CDS (Chromatography Data Systems)
- Descrizione:Dell™ mouse, bundled with Dell™ keyboard, DK
- Seq:22
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.