Labels, hazard warning
Hazard labels: transportation vehicle, PIC 306-TRI 100-B7541, Poliestere, nero on giallo, 100×87 mm, 3 labels/card
Catalog # BRDY250219
Supplier: Brady
Jump to:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specifications
- Colore:nero on giallo
- Descrizione:Hazard labels: transportation vehicle
- Dimensioni:100×87 mm
- Materiale:Poliestere
- Seq:23
- Modello:PIC 306-TRI 100-B7541
- Confezione:3 labels/card