Accessori
Left side: 2 shelves, 1 perforated insert, 1 bottom collecting sump (V=21 l) stainless steel 1.4301
Right side: 4 pull-out shelves with plastic sump melamine / PP
Right side: 4 pull-out shelves with plastic sump melamine / PP
Catalog # MSGA30755
Supplier: asecos
Jump to:
Product Details & Documents
Specifications
- Descrizione:Left side: 2 shelves, 1 perforated insert, 1 bottom collecting sump (V=21 l) stainless steel 1.4301
Right side: 4 pull-out shelves with plastic sump melamine / PP