Accessori
Left side: 4 drawers, stainless steel 1.4301
Right side: 6 pull-out shelves with plastic sump melamine / PP
Right side: 6 pull-out shelves with plastic sump melamine / PP
Catalog # MSGA30757
Supplier: asecos
Jump to:
Product Details & Documents
Specifications
- Descrizione:Left side: 4 drawers, stainless steel 1.4301
Right side: 6 pull-out shelves with plastic sump melamine / PP