CF660 GOAT ANTI-RABBIT IGG(H+L) 2 MG/ML 1 * 1 mg
Catalog # 20053-1MG
Supplier: Biotium
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
Supplier: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
VWR®, Salviette per il viso
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
VWR®, Guanti in Nitrile
Supplier: VWR Collection
Strisce reattive pH, MQuant®, Supelco®
Supplier: Merck
VWR®, Pipette sierologiche, Standard Line
Supplier: VWR Collection
Guanti in nitrile, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, pipette di trasferimento, alte prestazioni
Supplier: VWR Collection
VWR® NITRILE LIGHT, Guanti in nitrile
Supplier: VWR Collection