ContactUsLinkComponent
Human BD-4 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human BD-4 in a sandwich ELISA format. Using the ELISA protocol described below, the recommended microplates, reagen 1 * 200 Tests
Codice catalogo PEPR900-M435
Nome del fornitore:  PeproTech, Inc.
Human BD-4 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human BD-4 in a sandwich ELISA format. Using the ELISA protocol described below, the recommended microplates, reagen 1 * 200 Tests
Codice catalogo PEPR900-M435
Nome del fornitore:  PeproTech, Inc.
Frequently Bought Together