Human IL-4 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-4 in a sandwich ELISA format. Using the ELISA protocol described below, the recommended microplates, reagen 1 * 200 Tests
PEPR900-M14
Frequently Bought Together