Cleanroom labels, matte
Removable cleanroom labels, matte finish, PP, Acrilico, bianco, 38×38 mm, 1000 labels/roll
Codice catalogo 817-0301
Nome del fornitore: Texwipe
Vai a
Product Details & Documents
Matte finish cleanroom labels provide excellent compatibility with cleanroom pens and thermal transfer inks. Available with two different adhesive levels to provide excellent adhesion to a variety of surfaces without leaving residue.
- Ideal for handwritten applications
Specifiche
- Colore:bianco
- Descrizione:Removable cleanroom labels, matte finish
- Dimensioni:38×38 mm
- Materiale:PP
- Seq:01
- Confezione:1000 labels/roll
- Adesivo:Acrilico