Orders LinkContactUsLinkComponent
Peqlab, PCR Tubes
PCR tubes with flat caps, trasparente, 0,5 ml, 4 bags of 250
  732-3207
   Produzione a basso impatto
Peqlab, PCR Tubes
  732-3207
   Produzione a basso impatto
Peqlab, PCR Tubes
Peqlab, PCR Tubes
PCR tubes with flat caps, trasparente, 0,5 ml, 4 bags of 250
SFr. 74.50
1 * 1.000 ST
These PCR tubes with caps are made of PP resin and are designed to fit most popular brands of thermal cyclers.
  • Ultra-thin walls for rapid heat transfer
  • Certified free from DNase, RNase and human DNA
  • Manufactured from polypropylene under strict quality control guidelines
Available with domed or flat caps.
  • Confezione:
    4 bags of 250
  • Capacità:
    0,5 ml
  • Colore:
    trasparente
  • Descrizione:
    PCR tubes with flat caps
  • Cat. No.:
    732-3207
  • Environmentally Preferable:
    Produzione a basso impatto
  • Certificazione ambientale:
    IRECS