Peqlab, PCR Tubes
PCR tubes with flat caps, trasparente, 0,5 ml, 4 bags of 250
732-3207
These PCR tubes with caps are made of PP resin and are designed to fit most popular brands of thermal cyclers.
- Ultra-thin walls for rapid heat transfer
- Certified free from DNase, RNase and human DNA
- Manufactured from polypropylene under strict quality control guidelines
Available with domed or flat caps.
- Confezione:4 bags of 250
- Capacità:0,5 ml
- Colore:trasparente
- Descrizione:PCR tubes with flat caps
- Cat. No.:732-3207
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:IRECS