Peqlab, PCR Tubes
PCR tubes with domed cap, trasparente, 0,2 ml, 2 bags of 500
Codice catalogo 732-3206
Nome del fornitore: VWR Collection
Peqlab, PCR Tubes
Codice catalogo 732-3206
Nome del fornitore: VWR Collection
Vai a
Product Details & Documents
These PCR tubes with caps are made of PP resin and are designed to fit most popular brands of thermal cyclers.
- Ultra-thin walls for rapid heat transfer
- Certified free from DNase, RNase and human DNA
- Manufactured from polypropylene under strict quality control guidelines
Available with domed or flat caps.
Specifiche
- Confezione:2 bags of 500
- Capacità:0,2 ml
- Colore:trasparente
- Descrizione:PCR tubes with domed cap
- Cat. No.:732-3206
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:IRECS