HPLC syringes
Autosampler syringe, 22, 250 µl, 50 mm, LCQ
Codice catalogo 555-8224
Nome del fornitore: Thermo Fisher Scientific
Vai a
Product Details & Documents
These HPLC syringes are standard fixed needle syringes for Rheodyne/Valco™ injectors.
- Easy and accurate
- Reproducible manual injections
Applications include industrial chromatography.
Specifiche
- Per:LCQ
- Gauge:22
- Volume:250 µl
- Descrizione:Autosampler syringe
- Lunghezza:50 mm
- Seq:06