Orders LinkContactUsLinkComponent
Accessori
Accessori
Left side: 3 shelves, 1 perforated insert, 1 bottom collecting sump (V=21 l) stainless steel 1.4301
Right side: 4 Pull-out shelves with plastic sump melamine / PP
Codice catalogo MSGA30338
Nome del fornitore:   asecos
Accessori
Codice catalogo MSGA30338
Nome del fornitore:  asecos
Accessori
Accessori
Left side: 3 shelves, 1 perforated insert, 1 bottom collecting sump (V=21 l) stainless steel 1.4301
Right side: 4 Pull-out shelves with plastic sump melamine / PP
Price
SFr. 1’450.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Specifiche
  • Descrizione:
    Left side: 3 shelves, 1 perforated insert, 1 bottom collecting sump (V=21 l) stainless steel 1.4301
    Right side: 4 Pull-out shelves with plastic sump melamine / PP
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.