Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Plunger cans

Plunger cans

Spray Can, 2 l, 160×305 mm
Codice catalogo MSGA23138
Nome del fornitore:   asecos
Plunger cans
Codice catalogo MSGA23138
Nome del fornitore:  asecos
Plunger cans
Plunger cans

Spray Can, 2 l, 160×305 mm

Price
SFr. 130.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents

These plunger cans are dedicated to the safe moistening of cloth, wipes and tissues.

  • Unused liquid flows back into the container
  • Spring mounted plate includes flame barrier
  • Shock resistant and explosion proof (PE models only)
  • Cans with protector ring (stainless steel models only)
Specifiche
  • Ø×A:
    160×305 mm
  • Capacità:
    2 l
  • Descrizione:
    Spray Can