Plunger cans
Plunger Can, 1 l, 180×200 mm
MSGA23135
: asecos
These plunger cans are dedicated to the safe moistening of cloth, wipes and tissues.
- Unused liquid flows back into the container
- Spring mounted plate includes flame barrier
- Shock resistant and explosion proof (PE models only)
- Cans with protector ring (stainless steel models only)
- Ø×A:180×200 mm
- Capacità:1 l
- Descrizione:Plunger Can
- Seq:04