Plunger cans
Plunger Can, 2,5 l, 180×290 mm
Codice catalogo MSGA23134
Nome del fornitore: asecos
Vai a
Product Details & Documents
These plunger cans are dedicated to the safe moistening of cloth, wipes and tissues.
- Unused liquid flows back into the container
- Spring mounted plate includes flame barrier
- Shock resistant and explosion proof (PE models only)
- Cans with protector ring (stainless steel models only)
Specifiche
- Ø×A:180×290 mm
- Capacità:2,5 l
- Descrizione:Plunger Can
- Seq:10