Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
PharmPrep® P sorbent

PharmPrep® P sorbent

PharmPrep® P 100 RP-18e, 20 µm, 1 kg
Catalog # 1.19996.1000
PharmPrep® P sorbent
Catalog # 1.19996.1000
CAS Number:  
PharmPrep® P sorbent

PharmPrep® P 100 RP-18e, 20 µm, 1 kg

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents

These particles have a perfect spherical shape and are available in two particle sizes: 10 and 20 μm. Together with a pore diameter of 100 Å (10 nm), these new sorbents fit perfectly into the polishing step of small peptides like insulin and other biopharmaceutical and pharmaceutical APIs like antibiotics and hormones. This highly porous silica is produced by spray drying.

Because Merck Millipore performs the entire manufacturing process, you benefit from consistent batch-to-batch purification, while ensuring superior quality standards and regulatory compliance.

Mean particle size: 10 µm, 20 µm

Specific pore volume: 0,8 to 0,9 ml/g

Specific surface area: 320 to 400 m²/g

Specifications

Specifications

  • Dimensioni particelle:
    20 µm
  • Conf:
    1 kg
  • Descrizione:
    PharmPrep® P 100 RP-18e

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.