Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

PharmPrep® P sorbent

PharmPrep® P SI 100, 10 µm, 1 kg
  1.19681.1000
PharmPrep® P sorbent
  1.19681.1000
 :  
PharmPrep® P sorbent

PharmPrep® P SI 100, 10 µm, 1 kg

 :  
 
 
   
1 * 1 kg
 

These particles have a perfect spherical shape and are available in two particle sizes: 10 and 20 μm. Together with a pore diameter of 100 Å (10 nm), these new sorbents fit perfectly into the polishing step of small peptides like insulin and other biopharmaceutical and pharmaceutical APIs like antibiotics and hormones. This highly porous silica is produced by spray drying.

Because Merck Millipore performs the entire manufacturing process, you benefit from consistent batch-to-batch purification, while ensuring superior quality standards and regulatory compliance.

Mean particle size: 10 µm, 20 µm

Specific pore volume: 0,8 to 0,9 ml/g

Specific surface area: 320 to 400 m²/g

 

 

  • Dimensioni particelle:
    10 µm
  • Conf:
    1 kg
  • Descrizione:
    PharmPrep® P SI 100

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.