MOLYBDATE HR (0-100 MG/L) REAGENT TESTS 1 * 50 pezzi
Catalog # PALIPM175
Supplier: Palintest
Frequently Bought Together
Strisce reattive, nitriti, MQuant™, Supelco®
Supplier: Merck
Sodio idrossido 50% in soluzione acquosa, TECHNICAL
Supplier: VWR Chemicals
Acido ossalico 0.005 mol/l (0.01 N), AVS TITRINORM soluzione volumetrica
Supplier: VWR Chemicals
Acido 1-pentansolfonico sale sodico ≥99%, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
Non-aqueous DPX mounting medium
Supplier: Merck
Strisce reattive, perossidi, MQuant™, Supelco®
Supplier: Merck
pH indicator papers, universal/special
Supplier: MACHEREY-NAGEL
Sodio idrossido ≥97.0%, in sferette, TECHNICAL
Supplier: VWR Chemicals
Test strips, ammonia
Supplier: MACHEREY-NAGEL
Eyewash bottles, Duo
Supplier: Plum Safety
Masterflex® Correlated Variable-Area Flowmeters with Valves, High-Flow, 65-mm, Avantor®
Supplier: Avantor Fluid Handling
Test kit, fotometrico, Solfito, test in cuvetta LCK
Supplier: Hach
Powder pillows, ammonia
Supplier: Hach
Test kit, fotometrico, Solfato, test in cuvetta LCK
Supplier: Hach