Orders LinkContactUsLinkComponent
Sweatshirt, Acode® Basecamp 1734
Sweatshirt, Acode® Basecamp 1734
rosso, S
  KWIN100225331S
Sweatshirt, Acode® Basecamp 1734
  KWIN100225331S
Sweatshirt, Acode® Basecamp 1734
Sweatshirt, Acode® Basecamp 1734
rosso, S
SFr. 41.00
1 * 1 ST
Sweatshirt made of 80% cotton and 20% polyester, brushed inside.
  • Round neck
  • Neck, collar, cuffs and hem in rib with elastane
Weight: 280 g/m²
  • Taglia dell'abbigliamento:
    S
  • Colore:
    rosso
  • Seq:
    27
  • Tessuto:
    80% cotton/20% polyester
  • Cat. No.:
    KWIN100225331S
  • Genere:
    Per l'uomo
  • Sleeve or leg length:
    Long
  • Risk area:
    Cold
  • Peso:
    280 g/m²
  • Modello:
    1734
  • Applicazioni per abiti:
    Industrial