Shell jacket, GEN-Y Cocona®
nero, XL, Regular
Codice catalogo KWIN100963940XL
Nome del fornitore: FRISTADS KANSAS
Vai a
Product Details & Documents
Breathable jacket made of 100% polyester with 3,5-layer Cocona® PU laminate fabric. The fabric is windproof and waterproof.
- Dot-reinforcement at shoulders, front and lower sleeves
- Fleece-lined collar, outer and inside storm flap over front zip
- Double front pockets with logo zip, two inside pockets
- Raglan sleeves, reflector piping at hem
- Detachable hood with snap fasteners
- Adjustable strap with reflector tape on sleeves
Weight: 119 g/m²
Documents
Specifiche
- Taglia dell'abbigliamento:XL
- Vestibilità del capo:Regular
- Colore:nero
- Seq:19
- Tessuto:Polyester
- Cat. No.:KWIN100963940XL
- Genere:Unisex
- Sleeve or leg length:Long
- Risk area:Rain (waterproof)
- Peso:119 g/m²
- Modello:Gen-Y
- Applicazioni per abiti:Outdoor
- Tipo di chiusura:Con chiusura a zip