Work jackets, Icon
blu scuro/royal blue, S
Codice catalogo KWIN100811576S
Nome del fornitore: FRISTADS KANSAS
Vai a
Product Details & Documents
Bi-coloured workers jacket made of 100% cotton.
- Contrast stitching
- Button front with storm flap
- Angle-cut sleeves, adjustable cuffs and waistband
- Broad back with yoke, reflector wings in side seams
- Two chest pockets with pouch and flap, mobile phone pocket with zip and storm flap, inside pocket
- Diagonal cut on front with integrated side pockets with internal pouch
Weight: 375 g/m²
Documents
Specifiche
- Taglia dell'abbigliamento:S
- Colore:blu scuro/royal blue
- Seq:02
- Tessuto:100% cotton
- Cat. No.:KWIN100811576S
- Genere:Unisex
- Sleeve or leg length:Long
- Peso:375 g/m²
- Applicazioni per abiti:Industrial
- Tipo di chiusura:Con bottoni
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.