Orders LinkContactUsLinkComponent
Thermoblock, SmartBlocks™
Thermoblock, SmartBlocks™
SmartBlock™ PCR 96, blocco termico per piastre per PCR da 96 pozzetti, coperchio incluso*
Codice catalogo 460-0234
   Produzione a basso impatto
Nome del fornitore:   EPPENDORF
Thermoblock, SmartBlocks™
Codice catalogo 460-0234
Nome del fornitore:  EPPENDORF
   Produzione a basso impatto
Thermoblock, SmartBlocks™
Thermoblock, SmartBlocks™
SmartBlock™ PCR 96, blocco termico per piastre per PCR da 96 pozzetti, coperchio incluso*
Price
SFr. 916.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Exchangeable in just seconds. Individually sensor-controlled SmartBlocks are your solution for maintaining accurate, uniform temperatures. A wide variety of these flexible thermoblocks for all common laboratory vessels support a broad range of applications - for use with Eppendorf ThermoMixer® C and ThermoStat C.
  • Optimised block design supports optimal temperature transfer to sample to achieve excellent results
  • Eppendorf SmartBlock-specific calibration by integrated sensor technology maximizes temperature accuracy and homogeneity
  • Insulated thermoblocks for ergonomic operation - you won't burn your fingers (PhysioCare Concept®)
  • Automatic block recognition for safe handling
  • Optimised block design ensures maximum temperature transfer to the sample
  • Eppendorf QuickRelease technology enables fast and simple block exchange – no need for tools
Every Eppendorf SmartBlock is individually inspected in production for temperature accuracy and includes a serial-number specific certificate for documentation purpose.
Documents
Specifiche
  • Descrizione:
    SmartBlock™ PCR 96, blocco termico per piastre per PCR da 96 pozzetti, coperchio incluso*
  • Seq:
    12
  • Cat. No.:
    460-0234
  • Environmentally Preferable:
    Produzione a basso impatto
  • Certificazione ambientale:
    Renewable Energy Certificate, ISO 14001