Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
(±)-α-Bromophenylacetic acid

(±)-α-Bromophenylacetic acid

Glass bottle for solids, 25 g
Codice catalogo ACTIAS22054G25
Marketsource
Nome del fornitore:   ACTIVATE SCIENTIFIC
(±)-α-Bromophenylacetic acid
Codice catalogo ACTIAS22054G25
Nome del fornitore:  ACTIVATE SCIENTIFIC
Marketsource
Numero CAS:  4870-65-9
(±)-α-Bromophenylacetic acid

Glass bottle for solids, 25 g

Numero CAS:  4870-65-9
Price
Richiedi prezzo
Log in per disponibilità
1 * 25 g
Product Details & Documents
Sinonimi:  α-Bromophenylacetic Acid, DL-α-Bromophenylacetic acid
  • Numero CAS:4870-65-9
  • Formula:C₈H₇BrO₂
  • Numero MDL:MFCD00004206
  • Molecular Weight:215.0440 g/mol
  • EINECS:225-477-4
  • UN:3261
  • ADR:8,II
Specifiche

Specifiche

  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    25 g

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.