Orders LinkContactUsLinkComponent
2-Acetylthiazole
Glass bottle for solids, 25 g
  ACTIAS21608G25
Marketsource
2-Acetylthiazole
  ACTIAS21608G25
Marketsource
 :  24295-03-2
2-Acetylthiazole
Glass bottle for solids, 25 g
 :  24295-03-2
1 * 25 g
 :  1-(Thiazol-2-yl)-1-ethanone
  •  :24295-03-2
  •  :C₅H₅NOS
  •  :MFCD00005324
  •  :127.17 g/mol
  •  :213 °C (1013 hPa)
  •  :65,5 °C
  •  :1,22 g/cm³ (20 °C)
  •  :Densità: 1,22 g/cm³ (20 °C)
  •  :246-134-5
  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    25 g