Mark-tubes for x-ray analysis
80 mm, 0,30 mm, 0,01 mm
HILG4001403
For x-ray analysis according to the method of Debye-Scherrer.
- Ø est:0,30 mm
- Spessore parete:0,01 mm
- Lunghezza:80 mm
- Seq:02
For x-ray analysis according to the method of Debye-Scherrer.