2-Methyl-8-quinolinol
Bottiglia di vetro, 100 g
Catalog # ACTIAS33511G100
Marketsource
Supplier: ACTIVATE SCIENTIFIC
2-Methyl-8-quinolinol
Catalog # ACTIAS33511G100
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 826-81-3
Jump to:
Product Details & Documents
Synonyms: 8-Hydroxy-2-methylquinoline, 8-Hydroxyquinaldine, 2-Methyloxine
- CAS Number:826-81-3
- Formula:C₁₀H₉NO
- MDL Number:MFCD00006765
- Molecular Weight:159.19 g/mol
- Boiling Pt:267 °C (1013 hPa)
- Melting Pt:70…72 °C
- Storage Temperature:Ambiente
- EINECS:212-562-6
- UN:3077
- ADR:9,III
Specifications
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:100 g
- Cat. No.:ACTIAS33511G100



