2-Methyl-8-quinolinol
Bottiglia di vetro, 10 g
Catalog # ACTIAS33511G10
Marketsource
Supplier: ACTIVATE SCIENTIFIC
2-Methyl-8-quinolinol
Catalog # ACTIAS33511G10
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 826-81-3
Jump to:
Product Details & Documents
Synonyms: 8-Hydroxy-2-methylquinoline, 8-Hydroxyquinaldine, 2-Methyloxine
- CAS Number:826-81-3
- Formula:C₁₀H₉NO
- MDL Number:MFCD00006765
- Molecular Weight:159.19 g/mol
- Boiling Pt:267 °C (1013 hPa)
- Melting Pt:70…72 °C
- Storage Temperature:Ambiente
- EINECS:212-562-6
- UN:3077
- ADR:9,III
Specifications
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



