5-Methoxy-2-nitrobenzonitrile
Glass bottle for solids, 5 g
Codice catalogo ACTIAS31724G5
Marketsource
Nome del fornitore: ACTIVATE SCIENTIFIC
5-Methoxy-2-nitrobenzonitrile
Codice catalogo ACTIAS31724G5
Nome del fornitore: ACTIVATE SCIENTIFIC
Marketsource
Numero CAS: 38469-84-0
Vai a
Product Details & Documents
- Numero CAS:38469-84-0
- Formula:C₈H₆N₂O₃
- Numero MDL:MFCD09056823
- Molecular Weight:178.1500 g/mol
Specifiche
Specifiche
- Tipo di conf.:Glass bottle for solids
- Conf:5 g
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



