3-Bromosalicylaldehyde
Glass bottle for solids, 1 g
Codice catalogo ACTIAS30567G1
Marketsource
Nome del fornitore: ACTIVATE SCIENTIFIC
3-Bromosalicylaldehyde
Codice catalogo ACTIAS30567G1
Nome del fornitore: ACTIVATE SCIENTIFIC
Marketsource
Numero CAS: 1829-34-1
Vai a
Product Details & Documents
Sinonimi: 3-Bromo-2-hydroxybenzaldehyde
- Numero CAS:1829-34-1
- Formula:C₇H₅BrO₂
- Numero MDL:MFCD00016587
- Molecular Weight:201.02 g/mol
Specifiche
Specifiche
- Tipo di conf.:Glass bottle for solids
- Conf:1 g
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



