3,4-Dichloropicolinic acid
Bottiglia di vetro, 5 g
Catalog # ACTIAS23673G5
Marketsource
Supplier: ACTIVATE SCIENTIFIC
3,4-Dichloropicolinic acid
Catalog # ACTIAS23673G5
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 959578-03-1
Jump to:
Product Details & Documents
- CAS Number:959578-03-1
- Formula:C₆H₃Cl₂NO₂
- MDL Number:MFCD01861980
Specifications
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:5 g



