Affinity chromatography columns, HiTrap™ KappaSelect
HiTrap™ KappaSelect, PP, 7×25 mm, 1 ml
Codice catalogo 17-5458-11
Nome del fornitore: Cytiva
Affinity chromatography columns, HiTrap™ KappaSelect
Codice catalogo 17-5458-11
Nome del fornitore: Cytiva
Vai a
Product Details & Documents
KappaSelect is an affinity medium designed for the purification of human Fab (kappa) fragments.
Antibody fragments, especially Fabs, are getting increased attention as potential biopharmaceuticals because they have some advantages over monoclonal antibodies (MAbs).
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Ulteriori dettagli sul prodotto
Specifiche
- Capacità:1 ml
- Descrizione:HiTrap™ KappaSelect
- Colonna:PP
- Dimensione delle sfere:7×25 mm
- Cat. No.:17-5458-11
- Stoccaggio:4 to 8°C, 20% Ethanol
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



