Razor blades, double edge prep
Double edge prep razor blades, individually wrapped, 0,10 mm, 38×19 mm
Codice catalogo PERS74-0002-0000
Nome del fornitore: PERSONNA MEDICAL
Razor blades, double edge prep
Codice catalogo PERS74-0002-0000
Nome del fornitore: PERSONNA MEDICAL
Vai a
Product Details & Documents
Personna® Double Edge Prep razor blades are produced using innovative technologies. The high quality stainless steel is carefully selected for optimal charateristics and corrosion resistance.
- Corrosion resistant
- Optimised coating and geometry for superior performance
- Reduced cut force
- Blades individually wrapped
Documents
PERS74-0002-0000_TDS_EN
Scheda dati
Specifiche
- Descrizione:Double edge prep razor blades, individually wrapped
- L×L:38×19 mm
- Seq:02
- Spessore:0,10 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



