Accessories for glove boxes, Protector®
Positive pressure conversion kit, incl. hose, two clamps and harness assembly, changes the built-in blower operation from standard negative pressure to positive pressure operation
Catalog # LBCP5441200
Supplier: LABCONCO
Jump to:
Product Details & Documents
Documents
LBCP5242200_TDS_EN
Datasheet
Specifications
- Descrizione:Positive pressure conversion kit, incl. hose, two clamps and harness assembly, changes the built-in blower operation from standard negative pressure to positive pressure operation
- Seq:19