Orders LinkContactUsLinkComponent
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Positive pressure conversion kit, incl. hose, two clamps and harness assembly, changes the built-in blower operation from standard negative pressure to positive pressure operation
Catalog # LBCP5441200
Supplier:   LABCONCO
Accessories for glove boxes, Protector®
Catalog # LBCP5441200
Supplier:  LABCONCO
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Positive pressure conversion kit, incl. hose, two clamps and harness assembly, changes the built-in blower operation from standard negative pressure to positive pressure operation
Price
SFr. 701.00
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Descrizione:
    Positive pressure conversion kit, incl. hose, two clamps and harness assembly, changes the built-in blower operation from standard negative pressure to positive pressure operation
  • Seq:
    19