Orders LinkContactUsLinkComponent
Accessories for glove boxes, Protector®
Replacement HEPA filter, 99,99% efficient on 0,03 micron particulates; for use with 5441400 or 5441403 kit
Codice catalogo LBCP5441401
Nome del fornitore:   LABCONCO
Accessories for glove boxes, Protector®
Codice catalogo LBCP5441401
Nome del fornitore:  LABCONCO
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Replacement HEPA filter, 99,99% efficient on 0,03 micron particulates; for use with 5441400 or 5441403 kit
Price
SFr. 363.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Specifiche
  • Descrizione:
    Replacement HEPA filter, 99,99% efficient on 0,03 micron particulates; for use with 5441400 or 5441403 kit