Orders LinkContactUsLinkComponent
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Moisture monitor (dew point measurements), 0 - 6000 ppm; incl. meter, probe, attachment plug, and calibration stand; for placement inside the glove box’s main chamber; 140×203×46 mm, 230 V, 50/60 Hz
Codice catalogo LBCP5442201
Nome del fornitore:   LABCONCO
Accessories for glove boxes, Protector®
Codice catalogo LBCP5442201
Nome del fornitore:  LABCONCO
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Moisture monitor (dew point measurements), 0 - 6000 ppm; incl. meter, probe, attachment plug, and calibration stand; for placement inside the glove box’s main chamber; 140×203×46 mm, 230 V, 50/60 Hz
Price
SFr. 6’750.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Documents
Specifiche
  • Descrizione:
    Moisture monitor (dew point measurements), 0 - 6000 ppm; incl. meter, probe, attachment plug, and calibration stand; for placement inside the glove box’s main chamber; 140×203×46 mm, 230 V, 50/60 Hz
  • Seq:
    15