Leflunomide
500 mg
Nome del fornitore: Thermo Fisher Scientific
Leflunomide
Codice catalogo J65917.ME
Nome del fornitore: Thermo Fisher Scientific
Numero CAS: 75706-12-6
Vai a
Product Details & Documents
CAS No.: 75706-12-6 Molecular Formula: C12H9F3N2O2 Formula Weight: 270.21 Storage Temperature: 2°C to 8°C Physical Form: Crystalline solid Appearance: White UN No.: UN2811 MDL No.: MFCD00867593
Applications: An immunosuppressive agent. Inhibits T and B cell proliferation.
Sinonimi: 5-Methyl-N-(4-(trifluoromethyl)phenyl)isoxazole-4-carboxamide, 5-Methyl-N-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxamide
- Numero CAS:75706-12-6
- Formula:C₁₂H₉F₃N₂O₂
- Numero MDL:MFCD00867593
- Molecular Weight:270.21 g/mol
- IndexMerck:14,5432
- ADR:6.1,III
Documents
Specifiche
Specifiche
- Conf:500 mg
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



