Orders LinkContactUsLinkComponent
L(-)-Glutatione (forma ridotta) ≥98%
Glass bottle for solids, 25 g
  J62166.14
L(-)-Glutatione (forma ridotta) ≥98%
  J62166.14
 :  70-18-8
L(-)-Glutatione (forma ridotta) ≥98%
L(-)-Glutatione (forma ridotta) ≥98%
Glass bottle for solids, 25 g
 :  70-18-8
SFr. 241.00
1 * 25 g
Purity: 98+% CAS No.: 70-18-8 Molecular Formula: C10H17N3O6S Formula Weight: 307.33 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Melting Point: ca 193° dec. Beilstein Reference: 1729812 Merck Reference: 14,4475 Optical Rotation: -17° (c=2 in water) MDL No.: MFCD00065939
A tripeptide used for reductive cleavage of disulfide bonds
 :  L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced
  •  :70-18-8
  •  :C₁₀H₁₇N₃O₆S
  •  :MFCD00065939
  •  :307.33 g/mol
  •  :14,4475
  •  :∼ 193 °C (dec.)
  •  :1,475 g/cm³ (20 °C)
  •  :Refrigerata
  •  :200-725-4
  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    25 g