L(-)-Glutatione (forma ridotta) ≥98%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
L(-)-Glutatione (forma ridotta) ≥98%
Catalog # J62166.06
Supplier: Thermo Fisher Scientific
CAS Number: 70-18-8
Jump to:
Product Details & Documents
Purity: 98+% CAS No.: 70-18-8 Molecular Formula: C10H17N3O6S Formula Weight: 307.33 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Melting Point: ca 193° dec. Beilstein Reference: 1729812 Merck Reference: 14,4475 Optical Rotation: -17° (c=2 in water) MDL No.: MFCD00065939
A tripeptide used for reductive cleavage of disulfide bonds
Synonyms: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced
- CAS Number:70-18-8
- Formula:C₁₀H₁₇N₃O₆S
- MDL Number:MFCD00065939
- Molecular Weight:307.33 g/mol
- Merck Index:14,4475
- Melting Pt:∼ 193 °C (dec.)
- Density:1,475 g/cm³ (20 °C)
- Storage Temperature:Refrigerata
- EINECS:200-725-4
Documents
Specifications
Specifications
- Tipo di conf.:Glass bottle for solids
- Conf:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



