Tetracycline
Glass bottle for solids, 100 g
Nome del fornitore: Thermo Fisher Scientific
Tetracycline
Codice catalogo J61714.22
Nome del fornitore: Thermo Fisher Scientific
Numero CAS: 60-54-8
Vai a
Product Details & Documents
CAS No.: 60-54-8 Molecular Formula: C22H24N2O8 Formula Weight: 444.43 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Appearance: Pale yellow Melting Point: 172-174° Solubility: Limited solubility in water. Soluble in 1M HCl with heating. Beilstein Reference: 2230417 Merck Reference: 14,9196 MDL No.: MFCD00151232
Can induce apoptosis in osteoclasts
- Numero CAS:60-54-8
- Formula:C₂₂H₂₄N₂O₈
- Numero MDL:MFCD00151232
- Molecular Weight:444.43 g/mol
- IndexMerck:14,9196
- Punto di fusione:172…174 °C
- EINECS:200-481-9
Specifiche
Specifiche
- Tipo di conf.:Glass bottle for solids
- Conf:100 g



