Tetracycline
Glass bottle for solids, 25 g
Nome del fornitore: Thermo Fisher Scientific
Tetracycline
Codice catalogo J61714.14
Nome del fornitore: Thermo Fisher Scientific
Numero CAS: 60-54-8
Vai a
Product Details & Documents
CAS No.: 60-54-8 Molecular Formula: C22H24N2O8 Formula Weight: 444.43 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Appearance: Pale yellow Melting Point: 172-174° Solubility: Limited solubility in water. Soluble in 1M HCl with heating. Beilstein Reference: 2230417 Merck Reference: 14,9196 MDL No.: MFCD00151232
Can induce apoptosis in osteoclasts
- Numero CAS:60-54-8
- Formula:C₂₂H₂₄N₂O₈
- Numero MDL:MFCD00151232
- Molecular Weight:444.43 g/mol
- IndexMerck:14,9196
- Punto di fusione:172…174 °C
- EINECS:200-481-9
Specifiche
Specifiche
- Tipo di conf.:Glass bottle for solids
- Conf:25 g
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



