Sudan black B
Glass bottle for solids, 100 g
CAS No.: 4197-25-5 Molecular Formula: C29H24N6 Formula Weight: 456.55 Physical Form: Powder Appearance: Dark brown to black Melting Point: 118-126° Beilstein Reference: 723248 Merck Reference: 14,8885 MDL No.: MFCD00006919
Histochemical stain for chromosomes
: CI 26150, Ceres black BN, Solvent Black 3, Fat Black HB
- :4197-25-5
- :C₂₉H₂₄N₆
- :MFCD00006919
- :456.55 g/mol
- :14,8885
- :118…126 °C
- :Ambiente
- :224-087-1
- Tipo di conf.:Glass bottle for solids
- Conf:100 g