Orders LinkContactUsLinkComponent
Sudan black B
Glass bottle for solids, 100 g
  J62268.22
Sudan black B
  J62268.22
 :  4197-25-5
Sudan black B
Sudan black B
Glass bottle for solids, 100 g
 :  4197-25-5
252.00 SFr.
1 * 100 g
CAS No.: 4197-25-5 Molecular Formula: C29H24N6 Formula Weight: 456.55 Physical Form: Powder Appearance: Dark brown to black Melting Point: 118-126° Beilstein Reference: 723248 Merck Reference: 14,8885 MDL No.: MFCD00006919
Histochemical stain for chromosomes
 :  CI 26150, Ceres black BN, Solvent Black 3, Fat Black HB
  •  :4197-25-5
  •  :C₂₉H₂₄N₆
  •  :MFCD00006919
  •  :456.55 g/mol
  •  :14,8885
  •  :118…126 °C
  •  :Ambiente
  •  :224-087-1
  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    100 g