Sudan black B
Glass bottle for solids, 10 g
Nome del fornitore: Thermo Fisher Scientific
Sudan black B
Codice catalogo J62268.09
Nome del fornitore: Thermo Fisher Scientific
Numero CAS: 4197-25-5
Vai a
Product Details & Documents
CAS No.: 4197-25-5 Molecular Formula: C29H24N6 Formula Weight: 456.55 Physical Form: Powder Appearance: Dark brown to black Melting Point: 118-126° Beilstein Reference: 723248 Merck Reference: 14,8885 MDL No.: MFCD00006919
Histochemical stain for chromosomes
Sinonimi: CI 26150, Ceres black BN, Solvent Black 3, Fat Black HB
- Numero CAS:4197-25-5
- Formula:C₂₉H₂₄N₆
- Numero MDL:MFCD00006919
- Molecular Weight:456.55 g/mol
- IndexMerck:14,8885
- Punto di fusione:118…126 °C
- temperatura di conservazione:Ambiente
- EINECS:224-087-1
Documents
Specifiche
Specifiche
- Tipo di conf.:Glass bottle for solids
- Conf:10 g
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



