Orders LinkContactUsLinkComponent
Flasks, flat bottom with standard ground joint
Flasks, flat bottom with standard ground joint
250 ml, 24/29 NS
Codice catalogo NEUB0752-15-0250
Nome del fornitore:   NEUBERT VOLUME GLASSWAERE
Flasks, flat bottom with standard ground joint
Codice catalogo NEUB0752-15-0250
Nome del fornitore:  NEUBERT VOLUME GLASSWAERE
Flasks, flat bottom with standard ground joint
250 ml, 24/29 NS
Price
SFr. 72.20
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Borosilicate glass 3,3, amber.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Amber stained glass provides protection for materials sensitive to light.
Specifiche
  • Cono femmina:
    24/29 NS
  • Capacità:
    250 ml
  • Seq:
    03