Disposable cannulas
Disposable cannulas, G21, verde, 120 mm, +, 0,8×120 mm
Catalog # EHRS15884892
Supplier: EHRHARDT SOEHNE
Jump to:
Product Details & Documents
Disposable cannulas.
- Pyrogen-free, not toxic
- With lancet cut
- EO-sterilisation
Certificates
Search by Lot NumberSpecifications
- Ø×L:0,8×120 mm
- Gauge:G21
- Sterili:+
- Colore:verde
- Descrizione:Disposable cannulas
- Lunghezza:120 mm
- Seq:26