Bag cutters
Irradiated bag cutter, sterile, indivually packed
Codice catalogo 811-0165
Nome del fornitore: MICRONOVA
Vai a
Product Details & Documents
Plastic cutters for opening raw material packages.
- Automatically retractable safety blade guard
- No blade changes required - eliminates all loose blades
- Ergonomic handle
- Available gamma irradiated
Documents
Specifiche
- Descrizione:Irradiated bag cutter, sterile, indivually packed
- Seq:02