Caps
Cap, including EPDM O-ring, nuts, ferrules and colour-sleeve adapter, S 40, 3 ports with 1/4-28 threads for 1/8" or 1/16" tubing, PP
Codice catalogo JOURJR-S-14002
Nome del fornitore: VICI JOUR RESEARCH
Vai a
Product Details & Documents
These caps top bottles are available with two, three, or four colour-coded 1/4-28 thread for ¹/₈ or ¹/₁₆" O.Ø tubing.
Specifiche
- Filettatura:S 40
- Descrizione:Cap, including EPDM O-ring, nuts, ferrules and colour-sleeve adapter
- Materiale:PP
- Seq:09
- Connettore:3 ports with 1/4-28 threads for 1/8" or 1/16" tubing