Orders LinkContactUsLinkComponent
Shelving systems, solid, Super Erecta®
Shelving systems, solid, Super Erecta®
Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 610×1524 mm, 2460NFS, Acciaio inossidabile 304, 14,0 kg
Codice catalogo 139-0365
Nome del fornitore:   Metro International
Shelving systems, solid, Super Erecta®
Codice catalogo 139-0365
Nome del fornitore:  Metro International
Shelving systems, solid, Super Erecta®
Shelving systems, solid, Super Erecta®
Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 610×1524 mm, 2460NFS, Acciaio inossidabile 304, 14,0 kg
Price
SFr. 1’180.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Perfect for applications involving spillage. Provides barrier between floor and bottom shelf contents to deter contamination.
  • Shelf design features a 3,2 mm raised 'ship’s edge' on all four sides to contain spills
  • Louvered/embossed shelf allows air circulation
  • Available in several styles and materials for a variety of applications
  • Shelves are constructed with 18-gauge stainless steel (type 304) or galvanized materials
Standard stainless steel solid shelves (Type 304): with epoxy-coated cast corners address the majority of applications for solid, corrosion resistant shelves or work surfaces. Available in flat solid and louvered/embossed styles.
Galvanized shelves: with uncoated cast corners are ideal for applications requiring a solid shelving or work surface and minimum resistance to corrosion. Available in flat solid and louvered/embossed styles.
Premium Autoclave/Cart Washable Stainless Steel solid shelves (Type 304): feature an all-stainless construction to withstand high temperature, corrosive applications. Available in flat solid style.
Ordering information: Each individual shelf and post is sold separately. Accessories are also available.
Specifiche
  • Descrizione:
    Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 610×1524 mm, 2460NFS
  • Materiale:
    Acciaio inossidabile 304
  • Seq:
    62
  • Peso:
    14,0 kg
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Label holders, for 75×32 mm labels
    Label holders, for 75×32 mm labels
    METO9990P
    Log in per disponibilità
    1 * 1 ST - SFr. 8.90
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante mobile, SiteSelect™, cromato, altezza 1590 mm
    Montante mobile, SiteSelect™, cromato, altezza 1590 mm
    139-0268
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    Log in per disponibilità
    1 * 1 ST - SFr. 53.80
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante mobile, SiteSelect™, cromato, altezza 2454 mm
    Montante mobile, SiteSelect™, cromato, altezza 2454 mm
    139-0270
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    Log in per disponibilità
    1 * 1 ST - SFr. 74.80
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante mobile, SiteSelect™, in acciaio inox, altezza 2454 mm
    Montante mobile, SiteSelect™, in acciaio inox, altezza 2454 mm
    139-0277
    Scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale Super Erecta Pro™
    Log in per disponibilità
    1 * 1 ST - SFr. 202.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Slanted label holders, for 75×32 mm labels
    Slanted label holders, for 75×32 mm labels
    METO9990SL
    Log in per disponibilità
    1 * 1 ST - SFr. 8.90
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante mobile, SiteSelect™, in acciaio inox, altezza 1590 mm
    Montante mobile, SiteSelect™, in acciaio inox, altezza 1590 mm
    139-0275
    Scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale Super Erecta Pro™
    Log in per disponibilità
    1 * 1 ST - SFr. 149.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante fisso, SiteSelect™, cromato, altezza 2200 mm
    Montante fisso, SiteSelect™, cromato, altezza 2200 mm
    139-0255
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    Log in per disponibilità
    1 * 1 ST - SFr. 74.80
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante fisso, SiteSelect™, cromato, altezza 875 mm
    Montante fisso, SiteSelect™, cromato, altezza 875 mm
    139-0251
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    Log in per disponibilità
    1 * 1 ST - SFr. 45.70
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Decorator shelf inlays, black/white, 610×610 mm
    Decorator shelf inlays, black/white, 610×610 mm
    METO2424BWI
    Log in per disponibilità
    1 * 1 ST - SFr. 76.60
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Plastic split sleeves, black
    Plastic split sleeves, black
    METO9985
    Log in per disponibilità
    1 * 4 ST - SFr. 16.10
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Clear label holders,1520 mm, for 1395×32 mm labels
    Clear label holders,1520 mm, for 1395×32 mm labels
    METO9990CL5
    Log in per disponibilità
    1 * 1 ST - SFr. 45.10
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante mobile, SiteSelect™, cromato, altezza 1880 mm
    Montante mobile, SiteSelect™, cromato, altezza 1880 mm
    139-0269
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    Log in per disponibilità
    1 * 1 ST - SFr. 58.70
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Aluminum split sleeves with stainless rings
    Aluminum split sleeves with stainless rings
    METO9986S
    Log in per disponibilità
    1 * 1 Un. - SFr. 129.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante mobile, SiteSelect™, in acciaio inox, altezza 1880 mm
    Montante mobile, SiteSelect™, in acciaio inox, altezza 1880 mm
    139-0276
    Scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale Super Erecta Pro™
    Log in per disponibilità
    1 * 1 ST - SFr. 164.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante fisso, SiteSelect™, in acciaio inox, altezza 1590 mm
    Montante fisso, SiteSelect™, in acciaio inox, altezza 1590 mm
    139-0260
    Scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale fisso, scaffale Super Erecta Pro™, scaffale Super Erecta Pro™
    Log in per disponibilità
    1 * 1 ST - SFr. 149.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante fisso, SiteSelect™, metroseal 3, altezza 1590 mm
    Montante fisso, SiteSelect™, metroseal 3, altezza 1590 mm
    METO63PK3
    Scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta Pro™
    Log in per disponibilità
    1 * 1 ST - SFr. 63.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Enclosure panel, chrome, 313×1518 mm, for nominal post height 1600 mm
    Enclosure panel, chrome, 313×1518 mm, for nominal post height 1600 mm
    METOEP36C
    Log in per disponibilità
    1 * 1 ST - SFr. 143.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante fisso, SiteSelect™, cromato, altezza 1590 mm
    Montante fisso, SiteSelect™, cromato, altezza 1590 mm
    139-0253
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    Log in per disponibilità
    1 * 1 ST - SFr. 53.80
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante fisso, SiteSelect™, metroseal 3, altezza 1895 mm
    Montante fisso, SiteSelect™, metroseal 3, altezza 1895 mm
    METO74PK3
    Scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta Pro™
    Log in per disponibilità
    1 * 1 ST - SFr. 68.60
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Post mounting shelf supports, width 530 mm, double, chrome plated
    Post mounting shelf supports, width 530 mm, double, chrome plated
    METO2WS21C
    Log in per disponibilità
    1 * 1 ST - SFr. 128.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante fisso, SiteSelect™, cromato, altezza 1895 mm
    Montante fisso, SiteSelect™, cromato, altezza 1895 mm
    139-0254
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    Log in per disponibilità
    1 * 1 ST - SFr. 58.70
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Post mounting shelf supports, width 355 mm, double, chrome plated
    Post mounting shelf supports, width 355 mm, double, chrome plated
    METO2WS14C
    Log in per disponibilità
    1 * 1 ST - SFr. 122.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Post mounting shelf supports, width 530 mm, single, chrome plated
    Post mounting shelf supports, width 530 mm, single, chrome plated
    METO1WS21C
    Log in per disponibilità
    1 * 1 ST - SFr. 85.90
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Post mounting shelf supports, width 355 mm, single, chrome plated
    Post mounting shelf supports, width 355 mm, single, chrome plated
    METO1WS14C
    Log in per disponibilità
    1 * 1 ST - SFr. 82.80
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Storage baskets, chrome plated, 190×440×225 mm, 5 kg
    Storage baskets, chrome plated, 190×440×225 mm, 5 kg
    METOH212C
    Log in per disponibilità
    1 * 1 ST - SFr. 114.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Storage baskets, chrome plated, 127×345×180 mm, 4 kg
    Storage baskets, chrome plated, 127×345×180 mm, 4 kg
    METOH209C
    Log in per disponibilità
    1 * 1 ST - SFr. 81.60
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Enclosure panel, chrome, 464×1791 mm, for nominal post height 1800 mm
    Enclosure panel, chrome, 464×1791 mm, for nominal post height 1800 mm
    METOEP57C
    Log in per disponibilità
    1 * 1 ST - SFr. 179.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Additional tabs for rod, bag of 12
    Additional tabs for rod, bag of 12
    METO9084Z
    Log in per disponibilità
    1 * 1 Un. - SFr. 20.40
Informazioni sul prodotto
  • Codice catalogo
    Materiale
    Disponibilità
    Price
  • Clear shelf inlays for Super Erecta® shelving system, 914×355 mm
    Clear shelf inlays for Super Erecta® shelving system, 914×355 mm
    METO1436CI-4
    Plastica
    Log in per disponibilità
    1 * 1 Un. - SFr. 147.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Hanger rail fits on posts along the width or length of the unit 457 mm, fits to size shelf width: 914 mm. Optional hooks can hang from rail at any point.
    Hanger rail fits on posts along the width or length of the unit 457 mm, fits to size shelf width: 914 mm. Optional hooks can hang from rail at any point.
    METOH118C
    Log in per disponibilità
    1 * 1 ST - SFr. 43.30
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Aluminum split sleeves with zinc rings perfect for high temperature or conductive applications ; come with zinc retainer rings to secure the shelf in place (one pack required/shelf)
    Aluminum split sleeves with zinc rings perfect for high temperature or conductive applications ; come with zinc retainer rings to secure the shelf in place (one pack required/shelf)
    METO9986Z
    Log in per disponibilità
    1 * 1 Un. - SFr. 21.70
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Extension display hanger (152 mm): snaps on Super Erecta® shelf hanger rails and shelf frames, accepts standard carded merchandise
    Extension display hanger (152 mm): snaps on Super Erecta® shelf hanger rails and shelf frames, accepts standard carded merchandise
    METOHHK6C
    Log in per disponibilità
    1 * 1 ST - SFr. 22.90
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Split sleeves, plastic, white
    Split sleeves, plastic, white
    METO9985W
    Log in per disponibilità
    1 * 1 Un. - SFr. 16.10
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Slanted label holders, for 75×32 mm labels
    Slanted label holders, for 75×32 mm labels
    METO9990CL
    Log in per disponibilità
    1 * 1 ST - SFr. 8.90
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Clear label holders, plastic, fits 914 mm shelfs, 725×32 mm
    Clear label holders, plastic, fits 914 mm shelfs, 725×32 mm
    METO9990CL3
    Log in per disponibilità
    1 * 1 ST - SFr. 26.60
Informazioni sul prodotto
  • Codice catalogo
    Materiale
    Disponibilità
    Price
  • Clear shelf inlays for Super Erecta® shelving system, 914×530 mm
    Clear shelf inlays for Super Erecta® shelving system, 914×530 mm
    METO2136CI-4
    Plastica
    Log in per disponibilità
    1 * 1 Un. - SFr. 159.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Swaged post, stainless steel with aluminium cap, 1600 mm dimension
    Swaged post, stainless steel with aluminium cap, 1600 mm dimension
    METO63UPS-SW
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelving
    Log in per disponibilità
    1 * 1 ST - SFr. 164.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Stationary post, SiteSelect™, chrome plated, 2454 mm height
    Stationary post, SiteSelect™, chrome plated, 2454 mm height
    METO96P
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    Log in per disponibilità
    1 * 1 ST - SFr. 85.30
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Mobile post, SiteSelect™, stainless steel, 699 mm height
    Mobile post, SiteSelect™, stainless steel, 699 mm height
    139-0272
    Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
    Log in per disponibilità
    1 * 1 ST - SFr. 106.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Stationary post, SiteSelect™, stainless steel, 875 mm height
    Stationary post, SiteSelect™, stainless steel, 875 mm height
    139-0258
    Super Erecta Pro™ and Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
    Log in per disponibilità
    1 * 1 ST - SFr. 120.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Stationary post, SiteSelect™, stainless steel, 875 mm height
    Stationary post, SiteSelect™, stainless steel, 875 mm height
    139-0257
    Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
    Log in per disponibilità
    1 * 1 ST - SFr. 106.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Mobile post, SiteSelect™, chrome plated, 1385 mm height
    Mobile post, SiteSelect™, chrome plated, 1385 mm height
    139-0267
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    Log in per disponibilità
    1 * 1 ST - SFr. 51.30
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Mobile post, SiteSelect™, stainless steel, 857 mm height
    Mobile post, SiteSelect™, stainless steel, 857 mm height
    139-0273
    Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
    Log in per disponibilità
    1 * 1 ST - SFr. 120.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Stationary post, SiteSelect™, chrome plated, 1385 mm height
    Stationary post, SiteSelect™, chrome plated, 1385 mm height
    139-0252
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelving
    Log in per disponibilità
    1 * 1 ST - SFr. 51.30
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Stationary post, SiteSelect™, stainless steel, 2200 mm height
    Stationary post, SiteSelect™, stainless steel, 2200 mm height
    139-0262
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    Log in per disponibilità
    1 * 1 ST - SFr. 193.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Mobile post, SiteSelect™, chrome plated, 191 mm height
    Mobile post, SiteSelect™, chrome plated, 191 mm height
    139-0263
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    Log in per disponibilità
    1 * 1 ST - SFr. 31.50
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Stationary post, SiteSelect™, chrome plated, 191 mm height
    Stationary post, SiteSelect™, chrome plated, 191 mm height
    139-0248
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    Log in per disponibilità
    1 * 1 ST - SFr. 31.50
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Mobile post, SiteSelect™, stainless steel, 1385 mm height
    Mobile post, SiteSelect™, stainless steel, 1385 mm height
    139-0274
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelving
    Log in per disponibilità
    1 * 1 ST - SFr. 131.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Mobile post, SiteSelect™, stainless steel, 370 mm height
    Mobile post, SiteSelect™, stainless steel, 370 mm height
    139-0271
    Super Erecta Pro™, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® solid shelving
    Log in per disponibilità
    1 * 1 ST - SFr. 79.70
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Mobile post, SiteSelect™, chrome plated, 370 mm height
    Mobile post, SiteSelect™, chrome plated, 370 mm height
    139-0264
    Super Erecta Pro™, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® solid shelving
    Log in per disponibilità
    1 * 1 ST - SFr. 35.20
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Stationary post, SiteSelect™, stainless steel, 370 mm height
    Stationary post, SiteSelect™, stainless steel, 370 mm height
    139-0256
    Super Erecta Pro™, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® solid shelving
    Log in per disponibilità
    1 * 1 ST - SFr. 79.70
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante fisso, SiteSelect™, in acciaio inox, altezza 1895 mm
    Montante fisso, SiteSelect™, in acciaio inox, altezza 1895 mm
    139-0261
    Scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale fisso, scaffale Super Erecta Pro™
    Log in per disponibilità
    1 * 1 ST - SFr. 164.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Post clamps, joins units together for maximum strength, zinc-plated
    Post clamps, joins units together for maximum strength, zinc-plated
    METO9994Z
    Log in per disponibilità
    1 * 1 ST - SFr. 15.50
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante mobile, SiteSelect™, metroseal 3, altezza 1880 mm
    Montante mobile, SiteSelect™, metroseal 3, altezza 1880 mm
    METO74UPK3
    Scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta Pro™
    Log in per disponibilità
    1 * 1 ST - SFr. 68.60
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante mobile, SiteSelect™, cromato, altezza 699 mm
    Montante mobile, SiteSelect™, cromato, altezza 699 mm
    139-0265
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    Log in per disponibilità
    1 * 1 ST - SFr. 42.70
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Stationary post, SiteSelect™, stainless steel, 1385 mm height
    Stationary post, SiteSelect™, stainless steel, 1385 mm height
    139-0259
    Super Erecta Pro™ and Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
    Log in per disponibilità
    1 * 1 ST - SFr. 131.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Montante fisso, SiteSelect™, cromato, altezza 699 mm
    Montante fisso, SiteSelect™, cromato, altezza 699 mm
    139-0250
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    Log in per disponibilità
    1 * 1 ST - SFr. 42.70
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Heavy duty aluminum split sleeves with zinc rings
    Heavy duty aluminum split sleeves with zinc rings
    METO9986HZ
    Log in per disponibilità
    1 * 1 ST - SFr. 56.90